MilliporeSigma
  • Synergistic action of N-nitrosodialkylamines and near-UV in the induction of chromosome aberrations in Chinese hamster lung fibroblasts in vitro.
Synergistic action of N-nitrosodialkylamines and near-UV in the induction of chromosome aberrations in Chinese hamster lung fibroblasts in vitro.
Mutation research (1995-12-01)
Y Yamashita, N Sumi, S Arimoto, H Hayatsu
ABSTRACT
N-Nitrosodialkylamines are promutagens and proclastogens, requiring metabolic activation for their actions. Previously, we showed that direct-acting bacterial mutagens can be formed from N-nitrosodialkylamines on exposure to near-UV. We have now found that N-nitrosodialkylamines with near-UV irradiation are clastogenic to Chinese hamster lung cells. When the cells in culture were irradiated with near-UV for 3 h in the presence of N-nitrosodimethylamine (NDMA), N-nitrosodiethylamine (NDEA), N-nitrosopyrrolidine (NPYR), N-nitrosopiperidine (NPIP) or N-nitrosomorpholine (NMOR), and then further incubated for a total period of 24 h with the N-nitrosodialkylamines, chromosome aberrations were induced. Neither the N-nitrosodialkylamine nor near-UV alone were clastogenic. Severe clastogenicity (> 50% of cells examined showing aberrations) was observed for 0.5 mM NDEA, NPYR and NPIP. The order of the clastogenic activity was NDEA, NPYR > NPIP, NDMA > NMOR. This order differed from that of bacterial mutagenicity previously reported for these N-nitrosodialkylamines plus near-UV, in which NMOR gave the strongest activity. The chromosome aberrations induced by the NPYR and NDEA plus near-UV in CHL-cells were inhibited by superoxide dismutase, glutathione and L-cysteine. Dimethylsulfoxide or D-mannitol, scavengers of hydroxy radical, and L-histidine, a scavenger of single oxygen, were ineffective. These results suggest that superoxide formed by a synergistic action of an N-nitrosodialkylamine and near-UV is the cause of the chromosome aberrations observed, an assumption consistent with the known ability of superoxide to cleave DNA.
Product Description
Supelco
1-Nitrosopiperidine solution, certified reference material, 5000 μg/mL in methanol
Pricing and availability is not currently available.