MilliporeSigma
  • Activators of epithelial Na+ channels inhibit cytosolic feedback control. Evidence for the existence of a G protein-coupled receptor for cytosolic Na+.
Activators of epithelial Na+ channels inhibit cytosolic feedback control. Evidence for the existence of a G protein-coupled receptor for cytosolic Na+.
The Journal of membrane biology (1998-05-16)
P Komwatana, A Dinudom, J A Young, D I Cook
ABSTRACT
We have previously shown that epithelial Na+ channels in mouse mandibular gland duct cells are controlled by cytosolic Na+ and Cl-, acting, respectively, via Go and Gi proteins. Since we found no evidence for control of epithelial Na+ channels by extracellular Na+ ([Na+]o), our findings conflicted with the long-held belief that Na+ channel activators, such as sulfhydryl reagents, like para-chloromercuriphenylsulfonate (PCMPS), and amiloride analogues, like benzimidazolylguanidinium (BIG) and 5-N-dimethylamiloride (DMA), induce their effects by blocking an extracellular channel site which otherwise inhibits channel activity in response to increasing [Na+]o. Instead, we now show that PCMPS acts by rendering epithelial Na+ channels refractory to inhibition by activated G proteins, thereby eliminating the inhibitory effects of cytosolic Na+ and Cl- on Na+ channel activity. We also show that BIG, DMA, and amiloride itself, when applied from the cytosolic side of the plasma membrane, block feedback inhibition of Na+ channels by cytosolic Na+, while leaving inhibition by cytosolic Cl- unaffected. Since the inhibitory effects of BIG and amiloride are overcome by the inclusion of the activated alpha-subunit of Go in the pipette solution, we conclude that these agents act by blocking a previously unrecognized intracellular Na+ receptor.
Sigma-Aldrich
2-Guanidinobenzimidazole, 95%
Pricing and availability is not currently available.