MilliporeSigma
  • Trace iron determination in aminoisophthalic acid using differential-pulse cathodic stripping voltammetry at carbon paste electrodes.
Trace iron determination in aminoisophthalic acid using differential-pulse cathodic stripping voltammetry at carbon paste electrodes.
Journal of pharmaceutical and biomedical analysis (1998-10-20)
A Komersová, M Bartos, K Kalcher, K Vytras
ABSTRACT
Application of differential-pulse cathodic stripping voltammetry using a carbon paste electrode (consisting of carbon powder and liquid paraffin) have been investigated for trace determination of iron in 5-aminoisophthalic acid (AIPA). Samples were dissolved in 1 M HC1, pH was adjusted to 4-5 after addition of EDTA. Voltammetric measurements were performed after filtration. No sample decomposition (mineralization) was necessary. The method showed a good linearity between current and concentration from 3 x 10(-7) to 5 x 10(-5) mol dm-3 of iron, with a detection limit of 3 x 10(-7) mol dm-3 (resp. 1 ppm in solid AIPA). The results agreed well to those obtained by atomic absorption spectrometry (AAS) using electrothermic atomisation. For AAS measurement, however, microwave digestion of samples was necessary.
Sigma-Aldrich
5-Aminoisophthalic acid, 94%
Pricing and availability is not currently available.