MilliporeSigma
  • [Identification of the functional activity of synthetic polyamine analogues using a biotest system based on highly proliferating cultured human cells].
[Identification of the functional activity of synthetic polyamine analogues using a biotest system based on highly proliferating cultured human cells].
Prikladnaia biokhimiia i mikrobiologiia (2013-06-26)
K V Lisitskaia, N A Sokueva, Iu G Malysheva, A V Ivanov, S S Shishkin, S P Siatkin
ABSTRACT
A new biotest system was developed based on highly proliferating human cell cultures (lines LNCaP and PC-3). With the help of this system, two known synthetic polyamines--alpha-difluoromethylornithine (DFMO) and methylglioxalbis(guanylhydrason) (MGBG)--as well as four new synthetic analogues difenyl containing amines (DFCA-1-DFCA-4) with molecular weights of 725.5 (DFCA-1), 755.5 (DFCA-2), 655.5 (DFCA-3), and 681.5 Da (DFCA-4) were tested. In this biotest system, DFMO (0.1-400 microM) did not reveal functional activity, whereas for MGBG a cytotoxic effect was registered (100-200 microM). DFCA-1, DFCA-2, and DFCA-4 had a similar effect at concentrations of 10 microM and higher; DFCA-3, at a concentration of 50 microM and higher. Thus, DFCA-1 has a higher level of antiproliferating activity and may be considered as the most potent cytostatic agent.
Sigma-Aldrich
DL-α-Difluoromethylornithine hydrochloride hydrate, solid, ≥97% (NMR)
Pricing and availability is not currently available.