MilliporeSigma
  • Production in vitro by the cytochrome P450 CYP94A1 of major C18 cutin monomers and potential messengers in plant-pathogen interactions: enantioselectivity studies.
Production in vitro by the cytochrome P450 CYP94A1 of major C18 cutin monomers and potential messengers in plant-pathogen interactions: enantioselectivity studies.
The Biochemical journal (1999-08-05)
F Pinot, I Benveniste, J P Salaün, O Loreau, J P Noël, L Schreiber, F Durst
ABSTRACT
The major C(18) cutin monomers are 18-hydroxy-9,10-epoxystearic and 9,10,18-trihydroxystearic acids. These compounds are also known messengers in plant-pathogen interactions. We have previously shown that their common precursor 9,10-epoxystearic acid was formed by the epoxidation of oleic acid in Vicia sativa microsomes (Pinot, Salaün, Bosch, Lesot, Mioskowski and Durst (1992) Biochem. Biophys. Res. Commun. 184, 183-193). Here we determine the chirality of the epoxide produced as (9R,10S) and (9S,10R) in the ratio 90:10 respectively. We further show that microsomes from yeast expressing the cytochrome P450 CYP94A1 are capable of hydroxylating the methyl terminus of 9,10-epoxystearic and 9,10-dihydroxystearic acids in the presence of NADPH to form the corresponding 18-hydroxy derivatives. The reactions were not catalysed by microsomes from yeast transformed with a void plasmid or in absence of NADPH. After incubation of a synthetic racemic mixture of 9,10-epoxystearic acid with microsomes of yeast expressing CYP94A1, the chirality of the residual epoxide was shifted to 66:34 in favour of the (9S,10R) enantiomer. Both enantiomers were incubated separately and V(max)/K(m) values of 16 and 3.42 ml/min per nmol of P450 for (9R, 10S) and (9S,10R) respectively were determined, demonstrating that CYP94A1 is enantioselective for the (9R,10S) enantiomer, which is preferentially formed in V. sativa microsomes. Compared with the epoxide, the diol 9,10-dihydroxystearic acid was a much poorer substrate for the omega-hydroxylase, with a measured V(max)/K(m) of 0.33 ml/min per nmol of P450. Our results indicate that the activity of CYP94A1 is strongly influenced by the stereochemistry of the 9, 10-epoxide and the nature of substituents on carbons 9 and 10, with V(max)/K(m) values for epoxide>oleic acid>diol.