MilliporeSigma
  • De-novo biosynthesis of chlorinated aromatics by the white-rot fungus Bjerkandera sp. BOS55. Formation of 3-chloro-anisaldehyde from glucose.
De-novo biosynthesis of chlorinated aromatics by the white-rot fungus Bjerkandera sp. BOS55. Formation of 3-chloro-anisaldehyde from glucose.
FEBS letters (1992-07-06)
E de Jong, J A Field, J A Dings, J B Wijnberg, J A de Bont
ABSTRACT
The white-rot fungus Bjerkandera sp. BOS55 produced de-novo several aromatic metabolites. Besides veratryl alcohol and veratraldehyde, compounds which are known to be involved in the ligninolytic system of several other white-rot fungi, other metabolites were formed. These included anisaldehyde, 3-chloro-anisaldehyde and a yet unknown compound containing two chlorine atoms. Additionally GC/MS analysis revealed the production of small amounts of anisyl alcohol and 3-chloro-anisyl alcohol. After 14 days, the extracellular fluid of Bjerkandera BOS55 contained 100 microM veratraldehyde and 50 microM 3-chloro-anisaldehyde. This is the first report of de-novo biosynthesis of simple chlorinated aromatic compounds by a white-rot fungus. Anisaldehyde and 3-chloro-anisaldehyde were also produced by Bjerkandera adusta but not by Phanerochaete chrysosporium.
Sigma-Aldrich
3-Chloro-4-methoxybenzaldehyde, 97%
Pricing and availability is not currently available.