MilliporeSigma
Oral altretamine used as salvage therapy in recurrent ovarian cancer.
Gynecologic oncology (2004-01-31)
John K Chan, Vera Loizzi, Alberto Manetta, Michael L Berman
ABSTRACT
Altretamine has reported efficacy in the treatment of recurrent ovarian cancer following platinum-based therapy. This report presents the cases of two long-term survivors with recurrent ovarian cancer given oral altretamine. Two patients diagnosed with stage IIIC ovarian cancer underwent optimal cytoreductive surgery. Both women were subsequently treated with platinum-based chemotherapy. One had persistent cancer documented 2 months post therapy, while the other was disease-free for 22 months before recurring. Both received altretamine in a salvage setting. Each of these women achieved a prolonged response to third-line altretamine therapy, and one of whom was disease-free for 4 years and the other remains disease-free over 7 years following initiation of salvage therapy. Outpatient-administered oral altretamine can provide a prolonged disease-free interval with minimal toxicity.
Sigma-Aldrich
2,4,6-Tris(dimethylamino)-1,3,5-triazine, 96%
Pricing and availability is not currently available.