MilliporeSigma
  • Direct calculation of electron transfer parameters through constrained density functional theory.
Direct calculation of electron transfer parameters through constrained density functional theory.
The journal of physical chemistry. A (2006-07-21)
Qin Wu, Troy Van Voorhis
ABSTRACT
It is shown that constrained density functional theory (DFT) can be used to access diabatic potential energy surfaces in the Marcus theory of electron transfer, thus providing a means to directly calculate the driving force and the inner-sphere reorganization energy. We present in this report an analytic expression for the forces in constrained DFT and their implementation in geometry optimization, a prerequisite for the calculation of electron transfer parameters. The method is then applied to study the symmetric mixed-valence complex tetrathiafulvalene-diquinone radical anion, which is observed experimentally to be a Robin-Day class II compound but found by DFT to be in class III. Constrained DFT avoids this pitfall of over-delocalization and provides a way to find the charge-localized structure. In another application, driving forces and inner-sphere reorganization energies are calculated for the charge recombination (CR) reactions in formanilide-anthraquinone (FA-AQ) and ferrocene-formanilide-anthraquinone (Fc-FA-AQ). While the two compounds have similar reorganization energies, the driving force in FA-AQ is 1 eV larger than in Fc-FA-AQ, in agreement with experimental observations and supporting the experimental conclusion that the anomalously long-lived FA-AQ charge-separated state arises because the electron transfer is in the Marcus inverted region.
Sigma-Aldrich
Formanilide, 99%
Pricing and availability is not currently available.