MilliporeSigma
  • Solvent-enhanced diastereo- and regioselectivity in the Pd(II)-catalyzed synthesis of six- and eight-membered heterocycles via cis-aminopalladation.
Solvent-enhanced diastereo- and regioselectivity in the Pd(II)-catalyzed synthesis of six- and eight-membered heterocycles via cis-aminopalladation.
Chemistry (Weinheim an der Bergstrasse, Germany) (2009-06-25)
Arpád Balázs, Anasztázia Hetényi, Zsolt Szakonyi, Reijo Sillanpää, Ferenc Fülöp
ABSTRACT
The Pd(II)-catalyzed intramolecular oxidative cyclization of tosyl-protected cis- and trans-N-allyl-2-aminocyclohexanecarboxamides was examined, and efficient syntheses of cyclohexane-fused pyrimidin-4-ones and 1,5-diazocin-6-ones were developed. In the course of the research, a marked solvent effect was observed on both the regio- and diastereoselectivity. Additionally, a novel Pd(II)-mediated domino oxidation, oxidative amination reaction was discovered. Our experimental and theoretical findings suggest that the reactions proceed via a cis-aminopalladation mechanism.