MilliporeSigma
  • Development and validation of a sensitive ultra performance liquid chromatography tandem mass spectrometry method for the analysis of fentanyl and its major metabolite norfentanyl in urine and whole blood in forensic context.
Development and validation of a sensitive ultra performance liquid chromatography tandem mass spectrometry method for the analysis of fentanyl and its major metabolite norfentanyl in urine and whole blood in forensic context.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-06-15)
Ruth Verplaetse, Jan Tytgat
ABSTRACT
Fentanyl and its major metabolite norfentanyl often occur in low doses in biological samples. Therefore, a highly sensitive liquid chromatography tandem mass spectrometry (LC-MS/MS) method has been developed and fully validated. Sample preparation was performed on a mixed-mode cation exchange solid phase extraction (SPE) cartridge with an additional alkaline wash step to decrease matrix effects and thus increase sensitivity. Ionization of fentanyl and norfentanyl with electrospray ionization (ESI) was more efficient than atmospheric pressure chemical ionization (APCI). The use of a mobile phase of high pH resulted in higher ESI signals than the conventional low pH mobile phases. In the final method, gradient elution with 10 mM ammonium bicarbonate (pH 9) and methanol was performed. A comparison of columns with different internal diameter and/or smaller particles showed optimal resolution and sensitivity when an Acquity C18 column (1.7 microm, 2.1 mm x 50 mm) was used. Deuterium labeled internal standards were used, but with careful evaluation of their stability since loss of deuteriums was seen. With limits of detection of 0.25 pg/ml for fentanyl and 2.5 pg/ml for norfentanyl in urine and 5 pg/ml for fentanyl and norfentanyl in whole blood the presented method is highly appropriate for the analysis of fentanyl and norfentanyl in forensic urine and blood samples.
Product Description
Supelco
Norfentanyl oxalate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.