MilliporeSigma
  • Comparative toxicity of Euphorbia milii latex and synthetic molluscicides to Biomphalaria glabrata embryos.
Comparative toxicity of Euphorbia milii latex and synthetic molluscicides to Biomphalaria glabrata embryos.
Chemosphere (2010-07-03)
Eduardo C Oliveira-Filho, Barbara R Geraldino, Deise R Coelho, Rosângela R De-Carvalho, Francisco J R Paumgartten
ABSTRACT
Plant molluscicides have been regarded as possible alternatives to the costly and environmentally hazardous molluscicides currently available. This study was undertaken to compare the developmental toxicity of a plant molluscicide (Euphorbia milii latex, LAT) with that of three synthetic molluscicidal compounds. Biomphalaria glabrata egg masses (0-15 h after spawning) were exposed to molluscicides for 96 h and thereafter examined up to the 14th day after spawning. Embryo deaths, abnormal embryo development (malformations) and the day of hatching were recorded. Although exhibiting a weak ovicidal effect, LAT markedly impaired the development of snail embryos at concentrations 1000 microg L(-1) and produced anomalies (EC(50)=2040 microg L(-1)) such as abnormal shells, hydropic embryos, cephalic and non-specific malformations. Embryolethal potencies of molluscicides were as follows: triphenyltin hydroxide (TPTH; LC(50)=0.30 microg L(-1))>niclosamide (NCL; LC(50)=70 microg L(-1))>copper sulphate (CuSO(4); LC(50)=2190 microg L(-1)) > LAT (LC(50)=34030 microg L(-1)). A few malformations were recorded in embryos exposed to concentrations of TPTH within the range of lethal concentrations, while almost no anomalies were noted among those treated with NCL or CuSO(4). A hatching delay (hatching on day 10 after spawning or later) was observed among LAT-exposed embryos. The effects of NCL, TPTH and CuSO4 on hatching were to some extent masked by their marked embryolethality. The no-observed effect concentrations (NOEC) for embryotoxicity were as follows: TPTH, 0.1 microg L(-1); NCL, 25.0 microg L(-1); CuSO(4), 500.0 microg L(-1) and LAT, 500.0 microg L(-1). Results from this study suggest that, although LAT was not acutely embryolethal after a short-term exposure, it markedly disrupted snail development. The marked embryotoxicity of E. milii possibly contributes to its effectiveness as a molluscicide.
Sigma-Aldrich
Triphenyltin hydroxide
Pricing and availability is not currently available.
Supelco
Fentin hydroxide, PESTANAL®, analytical standard
Pricing and availability is not currently available.