MilliporeSigma
  • Topical sucralfate for pain after oral CO2 laser surgery: a prospective, randomized, controlled trial.
Topical sucralfate for pain after oral CO2 laser surgery: a prospective, randomized, controlled trial.
American journal of otolaryngology (2011-06-11)
Chau-Shiang Guo, Hui-Ching Chuang, Chih-Yen Chien
ABSTRACT
The aim of this study was to assess the effect of topical sucralfate on postoperative pain scores and other secondary outcomes including the frequency and duration of analgesic use and postoperative bleeding episodes after CO(2) laser treatment of oral leukoplakia. In this prospective trial, a total of 80 patients were randomized into the sucralfate group (n = 40) or the control group (n = 40). Postoperative pain scores, the frequency and duration of analgesic requirements, and postoperative wound bleeding episodes were compared between the 2 groups from the operative day to postoperative day 6. Patients in the sucralfate group experienced significantly less postoperative pain on postoperative days 1 and 2. Although there was no significant difference in frequency and duration of analgesic use between the 2 groups, a trend toward lower frequency and fewer days of analgesic use in the sucralfate group was observed. This study demonstrated the efficacy of topical sucralfate application in diminishing postoperative pain after CO(2) laser therapy for oral leukoplakia. Topical sucralfate can be considered a feasible adjuvant medication for the control of pain after CO(2) laser treatment of oral leukoplakia.
Sigma-Aldrich
Sucrose octasulfate–aluminum complex
Pricing and availability is not currently available.