MilliporeSigma
  • Chemical modification of the histidine residue in basic phospholipase A2 from the venom of Naja nigricollis.
Chemical modification of the histidine residue in basic phospholipase A2 from the venom of Naja nigricollis.
Biochimica et biophysica acta (1980-08-07)
C C Yang, K King
ABSTRACT
Phospholipase A2 from Naja nigricollis venom was separated into three fractions by chromatography on a column of CM-Sephadex C-25. The pI values of fractions CMS-5, CMS-6 and CMS-9 were determined to be 7.6, 8.3 and 10.6, respectively. Fraction CMS-9 was further purified on a DEAE-Sephacel column and the homogeneity was verified. The specific activity of CMS-9 was found to be 1300 units per mg and lethal toxicity 0.3 mg per kg mouse. The most basic and toxic fraction, CMS-9, was subjected to chemical modification with p-bromophenzcyl bromide. The enzyme lost both the enzyme activity and lethal toxicity, however, the antigenicity remained unchanged. Although both 8-anilinonaphthalenesulfonate and Ca2+ showed pronounced protection on the inactivation process, the mechanism of 8-anilinonaphthalene-sulfonate protection is different from that of Ca2+. Amino acid analysis showed that only one (His-47) out of three histidine residues was modified. Although both native and His-modified CMS-9 were perturbed by the presence of Ca2+, the modified enzyme lost the characteristic tryptophan blue shift suggesting that the modified enzyme is unable to exert a charge effect upon Ca2+ binding in the vicinity of the tryptophan group. Scatchard plots revealed only one type of binding sites for 8-anilinonaphthalenesulfonate in the presence of Ca2+. On the other hand, the modified enzyme lost the ability to bind 8-anilinonaphthalene. It is suggested tentatively that the hydrophobic pocket in which 8-anilinonaphthalenesulfonate is bound may be the site of the enzyme that interacts with phospholipid.