MilliporeSigma
  • Detection of intracellular Factor VIII protein in peripheral blood mononuclear cells by flow cytometry.
Detection of intracellular Factor VIII protein in peripheral blood mononuclear cells by flow cytometry.
BioMed research international (2013-04-05)
Gouri Shankar Pandey, Sandra C Tseng, Tom E Howard, Zuben E Sauna
ABSTRACT
Flow cytometry is widely used in cancer research for diagnosis, detection of minimal residual disease, as well as immune monitoring and profiling following immunotherapy. Detection of specific host proteins for diagnosis predominantly uses quantitative PCR and western blotting assays. In this study, we optimized a flow cytometry-based detection assay for Factor VIII protein in peripheral blood mononuclear cells (PBMCs). An indirect intracellular staining (ICS) method was standardized using monoclonal antibodies to different domains of human Factor VIII protein. The FVIII protein expression level was estimated by calculating the mean and median fluorescence intensities (MFI) values for each monoclonal antibody. ICS staining of transiently transfected cell lines supported the method's specificity. Intracellular FVIII protein expression was also detected by the monoclonal antibodies used in the study in PBMCs of five blood donors. In summary, our data suggest that intracellular FVIII detection in PBMCs of hemophilia A patients can be a rapid and reliable method to detect intracellular FVIII levels.
Product Description
Human coagulation factor VIII concentrate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.