MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Preparation and characterization of EDTAD-modified magnetic-Fe3O4 chitosan composite: application of comparative adsorption of dye wastewater with magnetic chitosan.
Preparation and characterization of EDTAD-modified magnetic-Fe3O4 chitosan composite: application of comparative adsorption of dye wastewater with magnetic chitosan.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-07-05)
Hongyu Yang, Yunchun Li, Steven Sai Hang Ho, Xiumei Tian, Yunxue Xia, Yaou Shen, Maojun Zhao, Guangtang Pan
ABSTRACT
Ethylenediaminetetraacetic dianhydride (EDTAD)-modified magnetic-Fe3O4 chitosan (EMC), prepared using the cross-link agent glutaraldehyde and chemicals Fe3O4, chitosan, and EDTAD, was used to compare the adsorption of methylene blue (MB) with magnetic chitosan (MC). The composite structure was confirmed by multiple characterization techniques, including scanning electron microscopy (SEM), X-ray powder diffraction, Fourier transform infrared spectroscopy (FTIR), and potentiometric titration methods. The characterization results suggest that Fe3O4 particles successfully bound on the surface of chitosan, and the EDTAD thoroughly modified the MC. Furthermore, EMC had more amino, carboxyl, and hydroxy groups than typical MC. Adsorption conditions, such as pH values, initial concentrations of MB, reaction temperature, and contact time were systematically examined. In comparison, the maximum adsorption capacity of EMC was approximately twice as much as that of MC. The recovery efficiency for EMC was >80% using 0.1 M HCl as an eluent solution. Therefore, the results reported herein indicate that EMC is very attractive and imply a practical application for dye wastewater treatment.
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitosan, medium molecular weight