MilliporeSigma
  • Indoxyl-beta-D-glucuronide, the primary emitter of low-level chemiluminescence in plasma of hemodialysis patients.
Indoxyl-beta-D-glucuronide, the primary emitter of low-level chemiluminescence in plasma of hemodialysis patients.
Clinical chemistry (1994-08-01)
S Agatsuma, H Sekino, T Nagoshi, H Watanabe
ABSTRACT
Characteristic light emission induced by the oxidation of hydroxyl radicals has been found in plasma of hemodialysis patients (Agatsuma et al., Clin Chem 1992;38:48-55). We purified a primary emitter, a chemiluminescent component peaking at 430 nm, by anion-exchange chromatography and reversed-phase HPLC. By using proton nuclear magnetic resonance and authentic indoxyl compounds, we determined the primary emitter to be indoxyl-beta-D-glucuronide. Absorption and fluorescence spectra of the purified sample coincided well with those of authentic indoxyl-beta-D-glucuronide, as did the peak in the chemiluminescence emission spectrum. Retention time of the purified sample on reversed-phase HPLC, measured by fluorescence, was also in accordance with that of indoxyl-beta-D-glucuronide. To our knowledge, this is the first identification of a primary emitter of low-level chemiluminescence from a biological source.
Sigma-Aldrich
Indoxyl β-D-glucuronide cyclohexylammonium salt
Pricing and availability is not currently available.