MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Human gingival fibroblasts rescue butyric acid-induced T-cell apoptosis.
Human gingival fibroblasts rescue butyric acid-induced T-cell apoptosis.
Infection and immunity (2002-04-16)
Tomoko Kurita-Ochiai, Kuniyasu Ochiai, Naoto Suzuki, Kichibee Otsuka, Kazuo Fukushima
ABSTRACT
We previously demonstrated that butyric acid, an extracellular metabolite from periodontopathic bacteria, induces cytotoxicity and apoptosis in murine thymocytes, splenic T cells, and human Jurkat T cells. In this study, we used a cell-to-cell interaction system to examine the contribution of gingival fibroblasts to the regulation of T-cell death induced by butyric acid. Butyric acid slightly suppressed fibroblast viability in a concentration-dependent fashion. However, DNA fragmentation assays indicated that butyric acid did not induce apoptosis for up to 21 h in human gingival fibroblasts (Gin 1, F41-G, and H. pulp cells). The culture supernatants were assayed for interleukin 1alpha (IL-1alpha), IL-1beta, IL-6, IL-8, IL-11, tumor necrosis factor alpha, and transforming growth factor beta, but only the IL-6, IL-8, and IL-11 levels were significantly increased by addition of butyric acid. Butyric acid- or Fas-induced Jurkat-cell apoptosis was attenuated when Jurkat cells were cocultured with either F41-G or Gin 1 cells that had been preincubated for 6 h with butyric acid. IL-8 slightly stimulated butyric acid- or Fas-induced Jurkat-cell apoptosis in a dose-dependent manner, although a low dose of IL-8 had a mildly inhibitory effect on apoptosis. In contrast, IL-6 and IL-11 significantly suppressed butyric acid- or Fas-induced apoptosis in a dose-dependent fashion. Furthermore, the addition of monoclonal antibodies against human IL-6 and IL-11 to cocultures of gingival fibroblasts and Jurkat cells partially eliminated T-cell recovery. These results suggest that the proinflammatory cytokines such as IL-6 and IL-11, produced in fibroblasts stimulated with butyric acid, are involved in the attenuation of T-cell apoptosis by gingival fibroblasts.
Product Description
Supelco
Butyric acid, analytical standard
Sigma-Aldrich
Butyric acid, SAJ special grade, ≥99.5%
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
Butyric acid, ≥99%, FG