MilliporeSigma
  • Drinking drivers in Sweden who consume denatured alcohol preparations: an analytical-toxicological study.
Drinking drivers in Sweden who consume denatured alcohol preparations: an analytical-toxicological study.
Journal of analytical toxicology (1989-07-01)
A W Jones, M Lund, E Andersson
ABSTRACT
In the course of analyzing blood samples from drunk drivers, several low molecular weight volatiles were occasionally identified in addition to ethanol on the gas chromatograms. Among 21, 153 blood specimens analyzed during 1986, 77 contained ethanol as well as other volatile agents at the following mean concentrations: ethanol 2090 mg/L (range 830-3410), methanol 49.6 mg/L (range 20-178), acetone 88.3 mg/L (range 12-307), 2-propanol 32.2 mg/L (range 4-99), 2-butanone 49.2 mg/L (range 5-144), and 2-butanol 23.2 mg/L (range 4-64). A technical alcohol widely available in Sweden, trade name T-red, contains 92% w/w ethanol, 2% w/w acetone, and 5% w/w 2-butanone. A red coloring agent and a substance to impart a bitter taste (bitrex) are added to deter consumption. The drinking drivers who consumed technical alcohol were on average older (43 years compared with 35 years) and had higher mean BAC (2090 mg/L compared with 1767 mg/L). Those who drank denatured alcohol were more often apprehended while driving small motorcycles (mopeds) than were control groups of DWI offenders. The use of technical alcohol for intoxication might reflect, at least in part, the high costs and restricted availability of conventional alcoholic beverages in Sweden.
Sigma-Aldrich
Ethyl alcohol, denatured, denatured, reagent grade
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
5α-Pregnane-3,20-dione (allo), progesterone metabolite
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Progesterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Butanone, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
2-Propanol, JIS special grade, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Butanone, FCC, FG
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)