MilliporeSigma
  • Acetylation of aromatic ethers using acetic anhydride over solid acid catalysts in a solvent-free system. Scope of the reaction for substituted ethers.
Acetylation of aromatic ethers using acetic anhydride over solid acid catalysts in a solvent-free system. Scope of the reaction for substituted ethers.
Organic & biomolecular chemistry (2003-08-21)
Keith Smith, Gamal A el-Hiti, Anthony J Jayne, Michael Butters
ABSTRACT
The acetylation of aryl ethers using acetic anhydride in the presence of zeolites under modest conditions in a solvent-free system gave the corresponding para-acetylated products in high yields. The zeolite can be recovered, regenerated and reused to give almost the same yield as that given when fresh zeolite is used.
Sigma-Aldrich
2-Chloroanisole, 98%
Pricing and availability is not currently available.