MilliporeSigma
  • Preparation of highly substituted gamma-lactam follicle stimulating hormone receptor agonists.
Preparation of highly substituted gamma-lactam follicle stimulating hormone receptor agonists.
Bioorganic & medicinal chemistry (2005-08-16)
Jeffrey C Pelletier, John Rogers, Jay Wrobel, M Claudia Perez, Emily S Shen
ABSTRACT
An unusual combination of Weinreb amidation and Mitsunobu lactam formation was used to prepare highly substituted gamma-lactam analogues of a thiazolidinone follicle stimulating hormone receptor agonist. The analogue synthesis was stereoselective and the final products were chemically stable. Biological properties of the target molecules were nearly identical to those of the lead compound.
Sigma-Aldrich
3-Aminobenzonitrile, 99%
Pricing and availability is not currently available.