MilliporeSigma
  • Photodissociation dynamics of halogenated thiophenes at 235 nm: a resonance enhanced multiphoton ionization-time-of-flight (REMPI-TOF) study.
Photodissociation dynamics of halogenated thiophenes at 235 nm: a resonance enhanced multiphoton ionization-time-of-flight (REMPI-TOF) study.
The journal of physical chemistry. A (2012-10-20)
Monali Kawade, Ankur Saha, Hari P Upadhyaya, Awadhesh Kumar, Prakash D Naik, P N Bajaj
ABSTRACT
The photodissociation dynamics of halogen-substituted thiophenes, namely, 2-chlorothiophene and 2-bromo-5-chlorothiophene, has been studied in a supersonic molecular beam around 235 nm, using resonance enhanced multiphoton ionization (REMPI) time-of-flight (TOF) technique, by detecting the nascent state of the primary halogen atoms. A single laser has been used for excitation of halothiophenes, as well as for the REMPI detection of photoproducts, namely, chlorine and bromine atoms, in their spin-orbit states X((2)P(3/2)) and X*((2)P(1/2)). We have determined the translational energy distribution, the recoil anisotropy parameter, β, and the spin-orbit branching ratio, for chlorine and bromine atom elimination channels. State-specific TOF profiles are converted into kinetic energy distributions, using a least-squares fitting method, taking into account the fragment anisotropies, β(ι). The TOF profiles for Cl, Cl*, Br, and Br* are found to be independent of laser polarization; i.e., the β is well characterized by a value of ~0.0, within the experimental uncertainties. For 2-chlorothiophene, we have observed two components for the Cl and only one component for the Cl* atom elimination channel in the translational energy distributions. The average translational energies for the fast and the slow components of the Cl channel are 3.0 ± 1.0 and 1.0 ± 0.5 kcal/mol, respectively. For Cl*, the average translational energy is 3.5 ± 1.0 kcal/mol. For 2-bromo-5-chlorothiophene, we have observed only one component for Cl, Cl*, Br, and Br* in the translational energy distributions. The average translational energies for the Cl and Cl* channels are 3.5 ± 1.0 and 5.0 ± 1.0 kcal/mol, respectively, whereas the average translational energies for the Br and Br* channels are 2.0 ± 1.0 and 3.5 ± 1.0 kcal/mol, respectively. The energy partitioning into the translational modes is interpreted with the help of various models, such as impulsive and statistical models. The ΔH(f)(298) value for 2-chlorothiophene has been estimated theoretically to be 23.5 kcal/mol.