MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The influence of stem design on critical squeaking friction with ceramic bearings.
The influence of stem design on critical squeaking friction with ceramic bearings.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2013-07-03)
Na Fan, Michael M Morlock, Nicholas E Bishop, Gerd Huber, Norbert Hoffmann, Michele Ciavarella, Guang X Chen, Arne Hothan, Florian Witt
ABSTRACT
Ceramic-on-ceramic hip joints have been reported to squeak, a phenomenon that may occur in compromised lubrication conditions. One factor related to the incidence of in vivo squeaking is the stem design. However, it has not yet been possible to relate stem design to squeaking in deteriorating lubrication conditions. The purpose of this study was to determine critical friction factors for different stem designs. A hip simulator was used to measure the friction factor of a ceramic bearing with different stem designs and gradually deteriorating lubrication represented by evaporation of a volatile fluid lubricant. The critical squeaking friction factor was measured at the onset of squeaking for each stem. Critical friction was higher for the long cobalt chrome (0.32 ± 0.02) and short titanium stems (0.39 ± 0.02) in comparison with a long titanium stem (0.29 ± 0.02). The onset of squeaking occurred at a friction factor lower than that measured for dry conditions, in which squeaking is usually investigated experimentally. The results suggest that shorter or heavier stems might limit the possibility of squeaking as lubrication deteriorates. The method developed can be used to investigate the influence of design parameters on squeaking probability.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, suitable for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Acetone, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, suitable for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%