MilliporeSigma
  • High throughput liquid chromatography-tandem mass spectrometry assay for mercapturic acids of acrolein and crotonaldehyde in cigarette smokers' urine.
High throughput liquid chromatography-tandem mass spectrometry assay for mercapturic acids of acrolein and crotonaldehyde in cigarette smokers' urine.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-08-13)
Steven G Carmella, Menglan Chen, Adam Zarth, Stephen S Hecht
ABSTRACT
3-Hydroxypropylmercapturic acid (3-HPMA) and 3-hydroxy-1-methylpropylmercapturic acid (HMPMA) are urinary metabolites of the toxicants acrolein and crotonaldehyde, respectively. Virtually all human urine samples contain these metabolites, resulting from the action of glutathione-S-transferases on acrolein and crotonaldehyde, which are lipid peroxidation products, environmental and dietary contaminants, and constituents of cigarette smoke. We have developed a high throughput liquid chromatography-tandem mass spectrometry method for quantitative analysis of 3-HPMA and HMPMA in large numbers of small urine samples, as would be required in molecular epidemiology and clinical studies relating levels of these metabolites to cancer risk. Solid-phase extraction on mixed mode reverse phase-anion exchange 96-well plates provided sufficient purification for LC-MS/MS analysis, which was performed by auto-injection using a 96-well format, and resulted in clean, readily interpretable chromatograms, with detection limits of 4.5pmol/mL urine for 3-HPMA and 3.5pmol/mL urine for HMPMA. Accuracy was 92% for 3-HPMA and 97% for HMPMA while inter-day precision was 9.1% (coefficient of variation) for 3-HPMA and 11.0% for HMPMA. The method was applied to more than 2600 urine samples from smokers; mean levels of 3-HPMA and HMPMA were 4800±5358 (S.D.)pmol/mL and 3302±3341pmol/mL, respectively.
Sigma-Aldrich
Crotonaldehyde, mixture of cis and trans, ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Crotonaldehyde, predominantly trans, ≥99%, contains 0.1-0.2% BHT as stabilizer, 1% H2O as stabilizer
Pricing and availability is not currently available.