MilliporeSigma
  • Treatment and outcomes in patients with primary cutaneous B-cell lymphoma: the BC Cancer Agency experience.
Treatment and outcomes in patients with primary cutaneous B-cell lymphoma: the BC Cancer Agency experience.
International journal of radiation oncology, biology, physics (2013-09-05)
Sarah N Hamilton, Elaine S Wai, King Tan, Cheryl Alexander, Randy D Gascoyne, Joseph M Connors
ABSTRACT
To review the treatment and outcomes of patients with primary cutaneous B-cell lymphoma (CBCL). Clinical characteristics, treatment, and outcomes were analyzed for all patients referred to our institution from 1981 through 2011 with primary CBCL without extracutaneous or distant nodal spread at diagnosis (n=136). Hematopathologists classified 99% of cases using the World Health Organization-European Organization for Research and Treatment of Cancer (WHO-EORTC) guidelines. Median age at diagnosis was 62 years. Classification was 18% diffuse large B-cell leg-type (DLBCL-leg), 32% follicle center (FCCL), 45% marginal zone (MZL), and 6% nonclassifiable (OTHER). Of the 111 subjects with indolent lymphoma (FCCL, MZL, OTHER), 79% received radiation alone (RT), 11% surgery alone, 3% chemotherapy alone, 4% chemotherapy followed by RT, and 3% observation. Following treatment, 29% of subjects relapsed. In-field recurrence occurred in 2% treated with RT and in 33% treated with surgery alone. Of the 25 subjects with DLBCL-leg, 52% received chemotherapy followed by RT, 24% chemotherapy, 20% RT, and 4% surgery alone. Seventy-nine percent received CHOP-type chemotherapy (cyclophosphamide, doxorubicin or epirubicin, vincristine, prednisone), 47% with rituximab added. Overall and disease-specific survival and time to progression at 5 years were 81%, 92%, and 69% for indolent and 26%, 61%, and 54% for DLBCL-leg, respectively. On Cox regression analysis of indolent subjects, RT was associated with better time to progression (P=.05). RT dose, chemo, age>60 y, and >1 lesion were not significantly associated with time to progression. For DLBCL-leg, disease-specific survival at 5 years was 100% for those receiving rituximab versus 67% for no rituximab (P=.13). This review demonstrates better outcomes for indolent histology compared with DLBCL-leg, validating the prognostic utility of the WHO-EORTC classification. In the indolent group, RT was associated with 98% local control. DLBCL-leg is a more aggressive disease; the excellent results in the rituximab group suggest it has an important role in management.
Sigma-Aldrich
Epirubicin hydrochloride, ≥90% (HPLC)
Pricing and availability is not currently available.
Epirubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.