MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effects of chitosan on xenograft models of melanoma in C57BL/6 mice and hepatoma formation in SCID mice.
Effects of chitosan on xenograft models of melanoma in C57BL/6 mice and hepatoma formation in SCID mice.
Anticancer research (2013-11-14)
Ming-Yang Yeh, Ming-Fang Wu, Hung-Sheng Shang, Jin-Biou Chang, Yung-Luen Shih, Yung-Liang Chen, Hsiao-Fang Hung, Hsu-Feng Lu, Chun Yeh, W Gibson Wood, Fang-Ming Hung, Jing-Gung Chung
ABSTRACT
According to the World Health Organization, Complementary and alternative medicine (CAM) is a comprehensive term referring to traditional medical treatments and various forms of indigenous medicines, also known as indigenous or folk medicine. Cancer patients often use CAM in the form of nutritional supplements, psychological techniques and natural medical approaches in the place of or in parallel to conventional medicine. The present study aimed to determine if Chitosan can inhibit lung metastasis and hepatoma formation, by studying xenograft of B16F10 melanoma cells in C57BL/6 mice and of Smmu 7721 cells in SCID mice, respectively. For the lung metastasis model, after a five-week treatment, the survival rates of B6 mice were 15% for the control group and 35%, 20%, 45% and 40% for the 320,000 kDa, 173,000 kDa, 86,000 kDa and 8,000 kDa molecular-weight treatment groups, respectively. Chitosan treatment dramatically increased lifespan and inhibited tumor metastasis especially in treatment groups of the low-molecular weight compound. For the hepatoma growth model, the size of the liver tumor mass was approximately >14 mm in the control group. In comparison to the control group, the tumor mass grew slowly with Chitosan treatment, especially at the low-molecular weight treatment group. Chitosan slowed-down the rate of tumor growth but did not inhibit tumor formation. Data presented herein demonstrate that Chitosan has anticancer effects and thus further study of the substance is warranted to examine for mechanisms of action and optimal dosage.
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous