MilliporeSigma
  • Synthesis of novel quinolone and quinoline-2-carboxylic acid (4-morpholin-4-yl-phenyl)amides: a late-stage diversification approach to potent 5HT1B antagonists.
Synthesis of novel quinolone and quinoline-2-carboxylic acid (4-morpholin-4-yl-phenyl)amides: a late-stage diversification approach to potent 5HT1B antagonists.
Bioorganic & medicinal chemistry (2006-11-28)
Carey L Horchler, John P McCauley, James E Hall, Dean H Snyder, W Craig Moore, Thomas J Hudzik, Marc J Chapdelaine
ABSTRACT
Multiparallel amenable syntheses of 6-methoxy-8-amino-4-oxo-1,4-dihydroquinoline-2-carboxylic acid-(4-morpholin-4-yl-phenyl)amides (I) and 4-amino-6-methoxy-8-(4-methyl-piperazin-1-yl)-quinoline-2-carboxylic acid (4-morpholin-4-yl-phenyl)amides (II) which facilitate late-stage diversification at the 8-position of (I) and at the 4- and 8-positions of (II) are described. The resulting novel series were determined to contain potent 5HT(1B) antagonists. Preliminary SAR data are presented.