- Synthesis and structural characterisation of lower rim halogenated pyrogallol[4]arenes: bi-layers and hexameric nano-capsules.
Synthesis and structural characterisation of lower rim halogenated pyrogallol[4]arenes: bi-layers and hexameric nano-capsules.
Chemical communications (Cambridge, England) (2006-09-14)
Scott J Dalgarno, Nicholas P Power, Jochen Antesberger, Robert M McKinlay, Jerry L Atwood
PMID16969463
ABSTRACT
Two lower rim halogenated pyrogallol[4]arenes have been synthesised and structurally characterised as either bi-layer or hexameric nano-capsule motifs depending upon chain length and functionalisation.