MilliporeSigma
  • Sequential injection spectrophotometric determination of etilefrine hydrochloride.
Sequential injection spectrophotometric determination of etilefrine hydrochloride.
Farmaco (Societa chimica italiana : 1989) (2004-12-16)
Negussie W Beyene, Jacobus F van Staden, Raluca I Stefan
ABSTRACT
A simple, fast, economical and automated sequential injection spectrophotometric method for the determination of etilefrine hydrochloride is developed. The method is based on the condensation reaction of etilefrine hydrochloride with 4-aminoantipyrine in the presence of alkaline potassium hexacyanoferrate and the absorbance of the colored product measured at 503 nm. Aspiration order, flow rate, reaction coil diameter, reaction coil length, concentration of 4-aminoantipyrine and potassium ferricyanide, as well as aspiration volume of reagents and sample has been optimized. Using these optimized parameters, a linear relationship between the relative peak height and concentration was obtained in the range 1-20 mg l(-1). The detection limit (as 3sigma value) was 0.1 mg l(-1) and precision was 2.7% and 1.5% at 1 and 2 mg l(-1), respectively. This method is superior over previously reported ones in terms of linear range, short analysis time, high sample throughput, excellent reagent economy and minimum waste generation.
Product Description
Etilefrine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.