MilliporeSigma
  • Naturally occurring antifungal agents against Zygosaccharomyces bailii and their synergism.
Naturally occurring antifungal agents against Zygosaccharomyces bailii and their synergism.
Journal of agricultural and food chemistry (2005-06-23)
Ken-Ichi Fujita, Isao Kubo
ABSTRACT
Polygodial was found to exhibit a fungicidal activity against a food spoilage yeast, Zygosaccharomyces bailii, with the minimum fungicidal concentration (MFC) of 50 microg/mL (0.17 mM). The time-kill curve study showed that polygodial was fungicidal at any growth stage. The primary action of polygodial comes from its ability to disrupt the native membrane-associated function of integral proteins as nonionic surface active agents (surfactants) followed by a decrease in plasma membrane fluidity. The fungicidal activity of polygodial was increased 128-fold in combination with a sublethal amount (equivalent of 1/2 MFC) of anethole and vice versa relative to the fungicidal activity of anethole. The fungicidal activity of sorbic acid was enhanced 512-fold in combination with 1/2 MFC of polygodial. Conversely, the fungicidal activity of polygodial was enhanced 128-fold in combination with 1/2 MFC of sorbic acid.
Sigma-Aldrich
Polygodial, ≥97% (HPLC)
Pricing and availability is not currently available.