MilliporeSigma
  • Gene expression analysis of the mechanisms whereby black cohosh inhibits human breast cancer cell growth.
Gene expression analysis of the mechanisms whereby black cohosh inhibits human breast cancer cell growth.
Anticancer research (2007-05-01)
Linda Saxe Einbond, Tao Su, Hsan-Au Wu, Richard Friedman, Xiaomei Wang, Bei Jiang, Timothy Hagan, Edward J Kennelly, Fredi Kronenberg, I Bernard Weinstein
ABSTRACT
Previous studies indicate that specific extracts and the pure triterpene glycoside actein obtained from black cohosh inhibit growth of human breast cancer cells. Our aim is to identify alterations in gene expression induced by treatment with a methanolic extract (MeOH) of black cohosh. We treated MDA-MB-453 human breast cancer cells with the MeOH extract at 40 microg/ml and collected RNA at 6 and 24 h; we confirmed the microarray results with real-time RT-PCR for 18 genes. At 6 h after treatment there was significant increase in expression of ER stress (GRP78), apoptotic (GDF15), lipid biosynthetic (INSIG1 and HSD17B7) and Phase 1 (CYP1A1) genes and, at 24 h, decrease in expression of cell cycle (HELLS and PLK4) genes. Since the MeOH extract activated genes that enhance apoptosis and repressed cell cycle genes, it may be useful in the prevention and therapy of breast cancer.
Product Description
USP
Actein, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.