MilliporeSigma
  • Efficacy of a simethicone, activated charcoal and magnesium oxide combination (Carbosymag®) in functional dyspepsia: results of a general practice-based randomized trial.
Efficacy of a simethicone, activated charcoal and magnesium oxide combination (Carbosymag®) in functional dyspepsia: results of a general practice-based randomized trial.
Clinics and research in hepatology and gastroenterology (2011-04-12)
Benoit Coffin, Claude Bortolloti, Odile Bourgeois, Luc Denicourt
ABSTRACT
A simethicone, activated charcoal and magnesium oxide combination (Carbosymag(®)) has been used for almost 20 years in functional dyspepsia, but there is limited scientific evidence of efficacy. We evaluated the efficacy of Carbosymag(®) in 18- to 49-year-old patients with functional dyspepsia. A total of 276 dyspeptic patients consulting a general practitioner and meeting the Rome III criteria were included in this prospective placebo-controlled study. Variations in overall and individual dyspeptic symptoms were evaluated after 1 month of treatment. At the end of the treatment period, overall dyspeptic symptom intensity was significantly lower in the Carbosymag(®) group (P=0.01). The intensity of post-prandial fullness, epigastric pain, epigastric burning and abdominal bloating was significantly reduced in the Carbosymag(®) group relative to the placebo group (P<0.05). The number need to treat to induce a 70-% decrease in overall dyspeptic symptoms by Carbosymag(®) was 7 (IC 95%: 4-32). A simethicone, activated charcoal and magnesium oxide combination (Carbosymag(®)) was significantly more effective than a placebo on overall symptom intensity in dyspeptic patients consulting a general practitioner.
Product Description
USP
Simethicone, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.