- [Prokinetic drugs in gastrointestinal motility. Their application in pediatrics].
[Prokinetic drugs in gastrointestinal motility. Their application in pediatrics].
Boletin medico del Hospital Infantil de Mexico (1989-12-01)
J A Ramírez-Mayans, M Rivera-Echegoyen, P Casaubón-Garcín, R J Michel
PMID2576370
ABSTRACT
In this article, the neurophysiology and neurobiochemistry of the digestive system are reviewed briefly. The pharmacology of the main gastrointestinal prokinetic drugs and their indications, side effects and dosages in pediatric patients are described.
Product Description
Homatropine methylbromide, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Homatropine methylbromide for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.