MilliporeSigma
Inhibition of eukaryotic DNA replication by geminin binding to Cdt1.
Science (New York, N.Y.) (2000-12-23)
J A Wohlschlegel, B T Dwyer, S K Dhar, C Cvetic, J C Walter, A Dutta
ABSTRACT
In all eukaryotic organisms, inappropriate firing of replication origins during the G2 phase of the cell cycle is suppressed by cyclin-dependent kinases. Multicellular eukaryotes contain a second putative inhibitor of re-replication called geminin. Geminin is believed to block binding of the mini-chromosome maintenance (MCM) complex to origins of replication, but the mechanism of this inhibition is unclear. Here we show that geminin interacts tightly with Cdt1, a recently identified replication initiation factor necessary for MCM loading. The inhibition of DNA replication by geminin that is observed in cell-free DNA replication extracts is reversed by the addition of excess Cdt1. In the normal cell cycle, Cdt1 is present only in G1 and S, whereas geminin is present in S and G2 phases of the cell cycle. Together, these results suggest that geminin inhibits inappropriate origin firing by targeting Cdt1.
Sigma-Aldrich
Anti-Geminin antibody, Mouse monoclonal, clone GEM-1, purified from hybridoma cell culture
Pricing and availability is not currently available.