MilliporeSigma
Imaging individual mRNA molecules using multiple singly labeled probes.
Nature methods (2008-09-23)
Arjun Raj, Patrick van den Bogaard, Scott A Rifkin, Alexander van Oudenaarden, Sanjay Tyagi
ABSTRACT
We describe a method for imaging individual mRNA molecules in fixed cells by probing each mRNA species with 48 or more short, singly labeled oligonucleotide probes. This makes each mRNA molecule visible as a computationally identifiable fluorescent spot by fluorescence microscopy. We demonstrate simultaneous detection of three mRNA species in single cells and mRNA detection in yeast, nematodes, fruit fly wing discs, and mammalian cell lines and neurons.