MilliporeSigma
  • A meta-analysis of genome-wide association studies identifies ORM1 as a novel gene controlling thrombin generation potential.
A meta-analysis of genome-wide association studies identifies ORM1 as a novel gene controlling thrombin generation potential.
Blood (2013-12-21)
Ares Rocanin-Arjo, William Cohen, Laure Carcaillon, Corinne Frère, Noémie Saut, Luc Letenneur, Martine Alhenc-Gelas, Anne-Marie Dupuy, Marion Bertrand, Marie-Christine Alessi, Marine Germain, Philipp S Wild, Tanja Zeller, Francois Cambien, Alison H Goodall, Philippe Amouyel, Pierre-Yves Scarabin, David-Alexandre Trégouët, Pierre-Emmanuel Morange
ABSTRACT
Thrombin, the major enzyme of the hemostatic system, is involved in biological processes associated with several human diseases. The capacity of a given individual to generate thrombin, called the thrombin generation potential (TGP), can be robustly measured in plasma and was shown to associate with thrombotic disorders. To investigate the genetic architecture underlying the interindividual TGP variability, we conducted a genome-wide association study in 2 discovery samples (N = 1967) phenotyped for 3 TGP biomarkers, the endogenous thrombin potential, the peak height, and the lag time, and replicated the main findings in 2 independent studies (N = 1254). We identified the ORM1 gene, coding for orosomucoid, as a novel locus associated with lag time variability, reflecting the initiation process of thrombin generation with a combined P value of P = 7.1 × 10(-15) for the lead single nucleotide polymorphism (SNP) (rs150611042). This SNP was also observed to associate with ORM1 expression in monocytes (P = 8.7 × 10(-10)) and macrophages (P = 3.2 × 10(-3)). In vitro functional experiments further demonstrated that supplementing normal plasma with increasing orosomucoid concentrations was associated with impaired thrombin generation. These results pave the way for novel mechanistic pathways and therapeutic perspectives in the etiology of thrombin-related disorders.
Sigma-Aldrich
Thrombin from bovine plasma, ≥60 NIH units/mg protein (biuret)
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, 40-500 NIH units/mg protein (biuret)
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, 600-2,000 NIH units/mg protein (biuret)
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)
Sigma-Aldrich
Thrombin from human plasma, 400-1000 NIH units/mg protein
Sigma-Aldrich
Thrombin human, BioUltra, recombinant, expressed in HEK 293 cells, aqueous solution, ≥95% (SDS-PAGE)
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, Suitable for routine use in the thrombin time test
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥1,000 NIH units/mg protein (E1%/280, 18.3)
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥2800 NIH units/mg protein (E1%/280, 18.3)
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, 1500-3500 NIH units/mg protein (E1%/280, 18.3), suitable for cell culture
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280, 18.3)