MilliporeSigma
  • Death domain-associated protein 6 (Daxx) selectively represses IL-6 transcription through histone deacetylase 1 (HDAC1)-mediated histone deacetylation in macrophages.
Death domain-associated protein 6 (Daxx) selectively represses IL-6 transcription through histone deacetylase 1 (HDAC1)-mediated histone deacetylation in macrophages.
The Journal of biological chemistry (2014-02-20)
Zhenyu Yao, Qian Zhang, Xia Li, Dezhi Zhao, Yiqi Liu, Kai Zhao, Yin Liu, Chunmei Wang, Minghong Jiang, Nan Li, Xuetao Cao
ABSTRACT
As a multifunctional nuclear protein, death domain-associated protein 6 (Daxx) regulates a wide range of biological processes, including cell apoptosis and gene transcription. However, the function of Daxx in innate immunity remains unclear. In our study, we show that Daxx is highly expressed in macrophages and localized in nucleus of macrophages. The expression of Daxx is significantly up-regulated by stimulation with TLR ligands LPS and poly(I:C). Silence of Daxx selectively represses IL-6 expression at transcription level in LPS-activated macrophages. Upon stimulation of LPS, Daxx specifically binds to the promoter of IL-6 and inhibits histone acetylation at IL-6 promoter region. Further mechanism analyses show that histone deacetylase 1 (HDAC1) interacts with Daxx and binds to the promoter of IL-6. Daxx silencing decreases the association of HDAC1 to IL-6 promoter. Therefore, our data reveal that Daxx selectively represses IL-6 transcription through HDAC1-mediated histone deacetylation in LPS-induced macrophages, acting as a negative regulator of IL-6 during innate immunity and potentially preventing inflammatory response because of overproduction of IL-6.