MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Glycine propionyl l-carnitine attenuates d-Galactosamine induced fulminant hepatic failure in wistar rats.
Glycine propionyl l-carnitine attenuates d-Galactosamine induced fulminant hepatic failure in wistar rats.
Chemico-biological interactions (2014-02-26)
Ajaz A Ganai, Sadaf Jahan, Amjid Ahad, M Z Abdin, Humaira Farooqi
ABSTRACT
Glycine propionyl l-carnitine (GPLC) is a propionyl ester of carnitine that includes an additional glycine component. The present study evaluated hepatoprotective effect of GPLC in d-Galactosamine (d-GalN) induced fulminant hepatic failure. Rats were intraperitonially administered d-GalN (700mg/kgBW). GPLC was given as a pre-treatment (35mg/kgBW/day) for 1month followed by a single dose of d-GalN on the 31st day. d-GalN administration resulted in increased mortality and serum ALT and AST activities. These increases were significantly attenuated by GPLC. d-GalN treatment increased hepatic lipid peroxidation and a decrease in reduced glutathione content was observed. GPLC pre-treatment significantly decreased lipid peroxidation and augmented the level of GSH. d-GalN increased the circulating level of TNF-α and ATM-Kinase and MAP-Kinase expression. GPLC supplementation inhibited the increase in serum TNF-α and ATM-Kinase and MAP-Kinase expression. d-GalN treatment increased the level of Bax and Caspase-3 m-RNA while as a decline was observed in Bcl2 m-RNA. GPLC prevented the increase in Caspase-3 and Bax m-RNA and at the same time augmented the expression of Bcl2 m-RNA. Our findings suggest that GPLC alleviates d-GalN induced liver injury by strengthening antioxidative defense system and reducing apoptotic signalling pathways.
Product Description
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Glutamic-Oxalacetic Transaminase from porcine heart, Type I, ammonium sulfate suspension, 200-500 units/mg protein
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, ammonium sulfate suspension, ≥75 units/mg protein
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Levocarnitine, European Pharmacopoeia (EP) Reference Standard
USP
Levocarnitine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Carnitine inner salt, synthetic, ≥98%
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)