MilliporeSigma
  • Degradation of 4-Chloro-3,5-Dimethylphenol by a Heterogeneous Fenton-Like Reaction Using Nanoscale Zero-Valent Iron Catalysts.
Degradation of 4-Chloro-3,5-Dimethylphenol by a Heterogeneous Fenton-Like Reaction Using Nanoscale Zero-Valent Iron Catalysts.
Environmental engineering science (2013-06-20)
Lejin Xu, Jianlong Wang
ABSTRACT
Degradation of 4-chloro-3,5-dimethylphenol (PCMX) by a heterogeneous Fenton-like process using nanoparticulate zero-valent iron (nZVI) and hydrogen peroxide (H
Sigma-Aldrich
4-Chloro-3,5-dimethylphenol, purum, ≥98.0% (T)
Pricing and availability is not currently available.