MilliporeSigma
  • Multiple bio-analytical methods to reveal possible molecular mechanisms of developmental toxicity in zebrafish embryos/larvae exposed to tris(2-butoxyethyl) phosphate.
Multiple bio-analytical methods to reveal possible molecular mechanisms of developmental toxicity in zebrafish embryos/larvae exposed to tris(2-butoxyethyl) phosphate.
Aquatic toxicology (Amsterdam, Netherlands) (2014-04-02)
Zhihua Han, Qiangwei Wang, Jie Fu, Hongshan Chen, Ye Zhao, Bingsheng Zhou, Zhiyuan Gong, Si Wei, Jun Li, Hongling Liu, Xiaowei Zhang, Chunsheng Liu, Hongxia Yu
ABSTRACT
The flame retardant tris(2-butoxyethyl) phosphate (TBEP) is a frequently detected contaminant in the environment, wildlife and human milk. The potentially toxic effects of TBEP and their underlying molecular mechanisms have not been elucidated. Here, zebrafish embryos were exposed to different concentrations of TBEP from 4 hours of post-fertilization (hpf) to 120 hpf, and effects on embryonic development and global protein expression patterns examined. Our results demonstrate that treatment with TBEP (0.8-100mg/L) causes a concentration- and time-dependent decrease in embryonic survival and the hatching percentage. The median lethal concentration was 10.7 mg/L at 120 hpf. Furthermore, exposure to 150 or 800 μg/L TBEP inhibited the degradation and utilization of vitellogenins and down-regulated the expression of proteins related to cation binding, and lipid transport, uptake and metabolism, accompanied by a decrease in heart rate and body length. Exposure to TBEP (150 or 800 μg/L) also decreased the expression of proteins involved in cell proliferation and DNA repair, and led to an increased number of apoptotic cells in the tail region. Collectively, our results suggest that exposure to TBEP causes toxicity in the developing zebrafish by inhibiting the degradation and utilization of nutrients from the mother and inducing apoptosis.
Sigma-Aldrich
Tris(2-butoxyethyl) phosphate, 94%
Pricing and availability is not currently available.