MilliporeSigma
  • Coupling between protein folding and allostery in the GroE chaperonin system.
Coupling between protein folding and allostery in the GroE chaperonin system.
Proceedings of the National Academy of Sciences of the United States of America (2000-03-04)
O Yifrach, A Horovitz
ABSTRACT
GroEL is an allosteric protein that facilitates protein folding in an ATP-dependent manner. Herein, the relationship between cooperative ATP binding by GroEL and the kinetics of GroE-assisted folding of two substrates with different GroES dependence, mouse dihydrofolate reductase (mDHFR) and mitochondrial malate dehydrogenase, is examined by using cooperativity mutants of GroEL. Strong intra-ring positive cooperativity in ATP binding by GroEL decreases the rate of GroEL-assisted mDHFR folding owing to a slow rate of the ATP-induced transition from the protein-acceptor state to the protein-release state. Inter-ring negative cooperativity in ATP binding by GroEL is found to affect the kinetic partitioning of mDHFR, but not of mitochondrial malate dehydrogenase, between folding in solution and folding in the cavity underneath GroES. Our results show that protein folding by this "two-stroke motor" is coupled to cooperative ATP binding.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate, ampule, ≥90% (HPLC)
Pricing and availability is not currently available.