MilliporeSigma
  • Protective roles of nitric oxide on seed germination and seedling growth of rice (Oryza sativa L.) under cadmium stress.
Protective roles of nitric oxide on seed germination and seedling growth of rice (Oryza sativa L.) under cadmium stress.
Ecotoxicology and environmental safety (2014-07-22)
Junyu He, Yanfang Ren, Xiulan Chen, Hui Chen
ABSTRACT
Nitric oxide (NO) is a bioactive molecule in plants which mediates a variety of physiological processes and responses to biotic and abiotic stresses including heavy metals. In the present study, the effects of exogenous NO donor sodium nitroprusside (SNP) on rice seed germination and seedlings growth were investigated under Cd stress and a possible mechanism was postulated. The results indicated that 100μM Cd significantly decreased rice seed germination index, vigor index, root and shoot lengths as well as fresh weight compared to control. Exogenous SNP dose-dependently attenuated the inhibition of rice seed germination and thereafter seedling growth caused by Cd. The promoting effect was most pronounced at 30μM SNP. Cd exposure caused oxidative stress by elevating hydrogen peroxide (H2O2) and malondialdehyde (MDA) contents in root and shoot of rice seedlings. 30μM SNP counteracted partly Cd toxicity by reducing the H2O2 and MDA contents of Cd-exposed seedlings. Meanwhile, application of SNP markedly stimulated the activities of superoxide dismutases (SOD), ascorbate peroxidases (APX), guaiacol peroxidase (POD) and catalases (CAT) compared with Cd treatment alone, thereby indicating the enhancement of the antioxidative capacity in the root and shoot under Cd stress. In addition, addition of 30μM SNP increased accumulation of proline in both root and shoot. The Cd accumulation in seedlings was significant reduced by SNP, implicating that the protective role of SNP was responsible for preventing Cd accumulation. However, the effects of SNP were reverted by addition of cPTIO, a NO scavenger, suggesting the protective roles of SNP might be related to the induction of NO. Furthermore, K3Fe(CN)6 and [Formula: see text] / [Formula: see text] had no similar roles as SNP. Based on these results, it can be concluded that SNP exerted an advantageous effect on alleviating the inhibitory effect of Cd on rice seed germination and seedling growth, which might interact with NO.
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Cadmium, wire reel, 0.1m, diameter 0.70mm, as drawn, 99.99+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, foil, light tested, 25x25mm, thickness 0.005mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 6mm disks, thickness 0.015mm, 99.7+%
Cadmium, rod, 50mm, diameter 40mm, 99.9%
Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.99+%
Cadmium, wire reel, 0.1m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.015mm, 99.7+%