MilliporeSigma
  • Clicking 1,2,4,5-tetrazine and cyclooctynes with tunable reaction rates.
Clicking 1,2,4,5-tetrazine and cyclooctynes with tunable reaction rates.
Chemical communications (Cambridge, England) (2011-12-14)
Weixuan Chen, Danzhu Wang, Chaofeng Dai, Donald Hamelberg, Binghe Wang
ABSTRACT
Substituted tetrazines have been found to undergo facile inverse electron demand Diels-Alder reactions with "tunable" reaction rates.