MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • 2-Methoxyestradiol synergizes with sorafenib to suppress hepatocellular carcinoma by simultaneously dysregulating hypoxia-inducible factor-1 and -2.
2-Methoxyestradiol synergizes with sorafenib to suppress hepatocellular carcinoma by simultaneously dysregulating hypoxia-inducible factor-1 and -2.
Cancer letters (2014-09-15)
Li Ma, Guangxin Li, Huaqiang Zhu, Xuesong Dong, Dali Zhao, Xian Jiang, Jie Li, Haiquan Qiao, Shaobin Ni, Xueying Sun
ABSTRACT
Sorafenib is the approved systemic drug of choice for advanced hepatocellular carcinoma (HCC), but has demonstrated limited benefits because of drug resistance. 2-Methoxyestradiol (2ME2) has been shown to be a promising anticancer drug against various types of cancers and acts by dysregulating hypoxia-inducible factor (HIF)-1. Hypoxic cancer cells are extremely resistant to therapies since they elicit strong survival ability due to the cellular adaptive response to hypoxia, which is controlled by HIF-1 and HIF-2. The present study has demonstrated that sorafenib downregulated the expression of HIF-1α, making the hypoxic response switch from HIF-1α- to HIF-2α-dependent pathways, resulting in upregulation of HIF-2α, which contributes to the insensitivity of hypoxic HCC cells to sorafenib. HIF-2α played a dominant role in regulating VEGF, thus sorafenib in turn increased the expression of VEGF (a downstream molecule of both HIF-1 and HIF-2) and cyclin D1 (a downstream molecule of HIF-2), but reduced the expression of LDHA (a downstream molecule of HIF-1), in hypoxic HCC cells. 2ME2 significantly reduced the expression of both HIF-1α and HIF-2α, and their downstream molecules, VEGF, LDHA and cyclin D1, rendering hypoxic HCC cells to increased sensitivity to 2ME2. 2ME2 also inhibited the nuclear translocation of HIF-1α and HIF-2α proteins, but had no effect on their mRNA expression. 2M2 synergized with sorafenib to suppress the proliferation and induction of apoptosis of HCC cells in vitro and in vivo, and inhibited tumoral angiogenesis. These results indicate that 2ME2 given in combination with sorafenib acts synergistically for treating HCC.
Product Description
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-Estradiol, ≥98%
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
2-Methoxyestradiol, powder
Sigma-Aldrich
β-Estradiol, analytical standard
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture