MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Phase II trial of carboplatin and pemetrexed as first-line chemotherapy for non-squamous non-small cell lung cancer, and correlation between the efficacy/toxicity and genetic polymorphisms associated with pemetrexed metabolism: Hokkaido Lung Cancer Clinical Study Group Trial (HOT) 0902.
Phase II trial of carboplatin and pemetrexed as first-line chemotherapy for non-squamous non-small cell lung cancer, and correlation between the efficacy/toxicity and genetic polymorphisms associated with pemetrexed metabolism: Hokkaido Lung Cancer Clinical Study Group Trial (HOT) 0902.
Cancer chemotherapy and pharmacology (2014-10-09)
Kenya Kanazawa, Hiroshi Yokouchi, Xintao Wang, Takashi Ishida, Yuka Fujita, Satoru Fujiuchi, Toshiyuki Harada, Masao Harada, Kei Takamura, Satoshi Oizumi, Ichiro Kinoshita, Yutaka Katsuura, Osamu Honjo, Tetsuya Kojima, Hirotoshi Dosaka-Akita, Hiroshi Isobe, Mitsuru Munakata, Masaharu Nishimura
ABSTRACT
This phase II study evaluated the response rate (RR) and safety of combination therapy with carboplatin (CBDCA) and pemetrexed (PEM) in Japanese patients with non-squamous non-small cell lung cancer (non-sq NSCLC). Further, the relationship between therapy efficacy/toxicity and genetic polymorphisms associated with PEM metabolism was analyzed. Forty-one patients received CBDCA at a dose targeting an area under the concentration-time curve of 5 mg/mL × min and PEM of 500 mg/m(2) on day 1 every 3 weeks. Single-nucleotide polymorphisms of the thymidylate synthase (TYMS) coding gene, the variable number of tandem repeat (VNTR) in the TYMS, and the methylenetetrahydrofolate reductase (MTHFR) coding gene were analyzed. The overall RR was 36.6 %. Median progression-free survival and median survival time were 4.7 months [95 % confidence interval (CI) 3.9-5.6 months] and 16.2 months (95 % CI 6.1-26.2 months), respectively. Epidermal growth factor receptor gene mutations were detected in 6 patients (14.6 %). The VNTR in the TYMS significantly correlated with anemia (p = 0.047) and thrombocytopenia (p = 0.038). This combination therapy was effective and tolerable in patients with advanced non-sq NSCLC. The VNTR in the TYMS appears to be a predictive factor for anemia and thrombocytopenia in patients treated with this regimen.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Carboplatin