MilliporeSigma
  • Activation of eNOS in endothelial cells exposed to ionizing radiation involves components of the DNA damage response pathway.
Activation of eNOS in endothelial cells exposed to ionizing radiation involves components of the DNA damage response pathway.
Biochemical and biophysical research communications (2014-12-17)
Masaki Nagane, Hironobu Yasui, Yuri Sakai, Tohru Yamamori, Koichi Niwa, Yuichi Hattori, Takashi Kondo, Osamu Inanami
ABSTRACT
In this study, the involvement of ataxia telangiectasia mutated (ATM) kinase and heat shock protein 90 (HSP90) in endothelial nitric oxide synthase (eNOS) activation was investigated in X-irradiated bovine aortic endothelial cells. The activity of nitric oxide synthase (NOS) and the phosphorylation of serine 1179 of eNOS (eNOS-Ser1179) were significantly increased in irradiated cells. The radiation-induced increases in NOS activity and eNOS-Ser1179 phosphorylation levels were significantly reduced by treatment with either an ATM inhibitor (Ku-60019) or an HSP90 inhibitor (geldanamycin). Geldanamycin was furthermore found to suppress the radiation-induced phosphorylation of ATM-Ser1181. Our results indicate that the radiation-induced eNOS activation in bovine aortic endothelial cells is regulated by ATM and HSP90.
Sigma-Aldrich
Geldanamycin from Streptomyces hygroscopicus, ≥98% (HPLC), powder
Pricing and availability is not currently available.