- 2-Chloro-pyridine-3-carboxamide.
2-Chloro-pyridine-3-carboxamide.
Acta crystallographica. Section E, Structure reports online (2009-01-01)
Yu-Peng Hua, Ying Xu, Xue-Hong Wei, Hong-Bo Tong
PMID21581971
ABSTRACT
In the crystal structure of the title compound, C(6)H(5)ClN(2)O, the dihedral angle between the pyridine ring and the carboxamine group is 63.88 (8)°. Inter-molecular N-H⋯N and N-H⋯O hydrogen bonds link the mol-ecules into a two-dimensional network.