MilliporeSigma
  • B7-H3 was highly expressed in human primary hepatocellular carcinoma and promoted tumor progression.
B7-H3 was highly expressed in human primary hepatocellular carcinoma and promoted tumor progression.
Cancer investigation (2014-05-03)
Feifei Wang, Guoyan Wang, Tongshen Liu, Guohua Yu, Guangbo Zhang, Xiying Luan
ABSTRACT
B7-H3 has been detected in different cancers and correlated to tumor progression and outcome in cancer patients. In this study, we investigated the expression of B7-H3 in tissues and cells of primary hepatocellular carcinoma (PHC) patients. The research showed that B7-H3 is aberrantly expressed in PHC tissues and cells, and its high expression on HepG2 cells significantly promotes cell proliferation, adhesion, migration, and invasion capacity; moreover, it inhibits the proliferation of CD8(+) T cells. Thus, B7-H3 may have a critical role in PHC and it may enhance tumor escape from the immune surveillance of CD8(+) T cells.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cd276
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human CD276
Pricing and availability is not currently available.