MilliporeSigma
  • Analysis and Exposure Assessment of Perchlorate in Korean Dairy Products with LC-MS/MS.
Analysis and Exposure Assessment of Perchlorate in Korean Dairy Products with LC-MS/MS.
Environmental health and toxicology (2011-11-30)
Sung-Hee Oh, Ji-Woo Lee, Pawlas Mandy, Jeong-Eun Oh
ABSTRACT
Perchlorate is an emerging contaminant that is found everywhere, including various foods. Perchlorate is known to disturb the production of thyroid hormones and leads to mental disorders in fetuses and infants, as well as metabolic problems in adults. In this study, we attempted to establish an LC-MS/MS method for measuring perchlorate in dairy products and used this developed method to investigate perchlorate levels in Korean milk and yogurt samples. The developed method of perchlorate analysis requires a shaker and 1% acetic acid/acetonitrile as the extracting solvent. Briefly, the samples were extracted and then centrifuged (4000 rpm, 1hour), and the supernatant was then passed through a Envi™ Carb SPE cartridge that had been prewashed sequentially with 6 mL of acetonitrile and 6 mL of 1% acetic acid in water. The final volume of the sample extract was adjusted to 40 mL with reagent water and the final sample was filtered through a 0.20-µm pore size PTFE (Polytetrafluoroethylene) syringe filter prior to LC-MS/MS. The average levels of perchlorate in milk and yogurt samples were 5.63 ± 3.49 µg/L and 3.65 ± 2.42 µg/L, respectively. The perchlorate levels observed in milk samples in this study were similar to those reported from China, Japan, and the United States. The exposure of Koreans to perchlorate through the consumption of dairy products was calculated based on the results of this study. For all age groups, the calculated exposure to perchlorate was below the reference of dose (0.7 µg/kg-day) proposed by the National Academy of Science, USA, but the perchlorate exposure of children was higher than that of adults. Therefore, further investigation of perchlorate in other food samples is needed to enable a more exact assessment of exposure of children to perchlorate.
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Pricing and availability is not currently available.