MilliporeSigma
  • Challenges in EGFRvIII detection in head and neck squamous cell carcinoma.
Challenges in EGFRvIII detection in head and neck squamous cell carcinoma.
PloS one (2015-02-07)
Sarah E Wheeler, Ann Marie Egloff, Lin Wang, C David James, Peter S Hammerman, Jennifer R Grandis
ABSTRACT
Head and neck squamous cell carcinoma (HNSCC) accounts for more than 5% of all cancers worldwide. The mortality rate of HNSCC has remained unchanged (approximately 50%) over the last few decades. Ubiquitous overexpression of wild type EGFR in many solid tumors has led to the development of EGFR targeted therapies. EGFR can be constitutively activated via several mechanisms including the truncated, EGFR variant III isoform (EGFRvIII). EGFRvIII lacks exons 2-7 and has been reported to be present in up to 20-40% of HNSCC. EGFRvIII has been shown to contribute to cetuximab resistance. The mechanisms leading to EGFRvIII expression in HNSCC are unknown. The present investigation was undertaken to determine the etiology of EGFRvIII in HNSCC. Fixed HNSCC and glioma tissues were analyzed by fluorescence in situ hybridization for EGFR amplification. DNA and RNA from fresh frozen specimens were used to determine the presence of EGFRvIII transcripts and the mechanisms of expression via PCR, RT-PCR and RNA sequencing. Unlike glioma, EGFRvIII expression in HNSCC did not correlate with EGFR amplification. We found evidence of genomic deletion of the exon 2-7 in 6 of 7 HNSCC cases examined, however, the presence of genomic deletion did not always result in mRNA expression of EGFRvIII. RNA sequencing with automated alignment did not identify EGFRvIII due to microhomology between intron 1 and exon 8. RNA sequencing analyzed by manual alignment methods did not correlate well with RT-PCR and PCR findings. These findings suggest that genomic deletion as well as additional regulatory mechanisms may contribute to EGFRvIII expression in HNSCC. Further, large scale automated alignment of sequencing are unlikely to identify EGFRvIII and an assay specifically designed to detect EGFRvIII may be necessary to detect this altered form of EGFR in HNSCC tumors.
Sigma-Aldrich
Epidermal Growth Factor Receptor human, lyophilized powder, ≥15,000 units/mg protein (Bradford)
Pricing and availability is not currently available.
Sigma-Aldrich
Epidermal Growth Factor Receptor human, buffered aqueous glycerol solution, 5,000-30,000 units/mg protein (Lowry)
Pricing and availability is not currently available.