MilliporeSigma
  • Mechanism of E-cadherin dimerization probed by NMR relaxation dispersion.
Mechanism of E-cadherin dimerization probed by NMR relaxation dispersion.
Proceedings of the National Academy of Sciences of the United States of America (2013-09-27)
Ying Li, Nicole L Altorelli, Fabiana Bahna, Barry Honig, Lawrence Shapiro, Arthur G Palmer
ABSTRACT
Epithelial cadherin (E-cadherin), a member of the classical cadherin family, mediates calcium-dependent homophilic cell-cell adhesion. Crystal structures of classical cadherins reveal an adhesive dimer interface featuring reciprocal exchange of N-terminal β-strands between two protomers. Previous work has identified a putative intermediate (called the "X-dimer") in the dimerization pathway of wild-type E-cadherin EC1-EC2 domains, based on crystal structures of mutants not capable of strand swapping and on deceleration of binding kinetics by mutations at the X-dimer interface. In the present work, NMR relaxation dispersion spectroscopy is used to directly observe and characterize intermediate states without the need to disrupt the strand-swapped binding interface by mutagenesis. The results indicate that E-cadherin forms strand-swapped dimers predominantly by a mechanism in which formation of a weak and short-lived X-dimer-like state precedes the conformational changes required for formation of the mature strand-swapped dimeric structure. Disruption of this intermediate state through mutation reduces both association and dissociation rates by factors of ~10(4), while minimally perturbing affinity. The X-dimer interface lowers the energy barrier associated with strand swapping and enables E-cadherins to form strand-swapped dimers at a rate consistent with residence times in adherens junctions.
Sigma-Aldrich
E-Cadherin/Fc Chimera from mouse, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder
Pricing and availability is not currently available.